missing translation for 'onlineSavingsMsg'
Learn More
Learn More
All-rac-alpha-Tocopherol, FCC, 96-102%, Spectrum™ Chemical
T1154, 10191-41-0, C29H50O2
$2613.31 - $2613.31
Chemical Identifiers
| CAS | 10191-41-0 |
|---|---|
| Molecular Formula | C29H50O2 |
| Molecular Weight (g/mol) | 430.72 |
| MDL Number | MFCD00072051 |
| InChI Key | GVJHHUAWPYXKBD-IEOSBIPESA-N |
| IUPAC Name | (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol |
| SMILES | CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
Description
Spectrum™ Chemical All-rac-alpha-Tocopherol, FCC is the synthetic form of Vitamin E. All-rac-alpha-Tocopherol is often found in fortified foods and nutritional supplements. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 10191-41-0 | |
| 430.72 | |
| GVJHHUAWPYXKBD-IEOSBIPESA-N | |
| CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 |
| C29H50O2 | |
| MFCD00072051 | |
| (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol |
Specifications
| 10191-41-0 | |
| Amber Glass Bottle | |
| MFCD00072051 | |
| GVJHHUAWPYXKBD-IEOSBIPESA-N | |
| (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol | |
| 96% to 102% |
| 100% | |
| C29H50O2 | |
| 1 L | |
| CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1 | |
| 430.72 | |
| FCC |