Learn More
Shawmut Protex™ All-Day-Comfort™ N95 Particulate Respirators
Made in the USA, molded-cup N95 respirator is easier to breathe through while offering a comfortable and secure fit for maximum protection and less eyeglass fogging. Meets ASTM F3407 Respirator Fit Capability standard to securely fit a wide range of face sizes and shapes.
$34.04 - $397.10
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
View More Specs
Products 2
Catalog Number Mfr. No. Model Size Strap Attachment Type Price Quantity  
Catalog Number Mfr. No. Model Size Strap Attachment Type Price Quantity  
17000342
View Documents
Shawmut Protex™
SR9520S12
SR9520S-12 Small Welded Two Strap Design
Pack of 20 Each for $34.04
Case of 12 Pack for $148.80
Only null left
17-448-400
View Documents
Shawmut Protex™
SR952012
SR9520-12 Medium/Large Two-Strap Design
Pack of 20 Each for $397.10
Case of 12 Pack for $4,084.44
Only null left
Description
Description
Made by Shawmut Corp™, this NIOSH-approved, disposable N95 particulate respirator meets NIOSH 42 CFR 84 N95 requirements for a minimum 95% filtration efficiency. Its patent-pending All-Day Comfort™ System includes a soft, non-abrasive inner shell precisely thermoformed for a superior fit; a lightweight, super-high-efficiency filter layer for easier breathing, a comfortable viscoelastic nose foam for a secure seal, and expanded force-curve elastic head straps that are engineered to fit a wider range of sizes.
  • Molded cup style N95 for superior seal and protection
  • 6
  • Better breathability and cooler, fresher air inside the cup with Proprietary Shawmut MB™ filtering technology
  • Meets ASTM F3407 Respirator Fit Capability Standard to fit widest range of face sizes and shapes
  • Provides respiratory protection by removing more than 95% of airborne particles larger than 0.3 microns
  • Secure fit and tight seal around user's face ensures the respirator will perform at N95 standards while being comfortable to wear for long periods
  • Less eyeglass fogging with unique viscoelastic nose foam and aluminum nose clip
  • Two-strap welded design for more secure fit
  • Ideal for sanding, cutting, sawing, yard work, painting, grinding or other heavy industries
  • Also appropriate for general masking and travel recommendations, as well as liquid or non-oil-based particles from sprays that do not also emit oil aerosols or vapors
  • Made in the USA
Specifications
Specifications
N - Not Resistant to Oil
Cup/Molded
N95
General Purpose Particulate Respirator
95%
Elastic
20 Pk.
No
NIOSH 42 CFR 84, N95
Single-use
Polypropylene
No
Yes
Adjustable Nose Clip
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Not made with natural rubber latex.