missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlkylFluor(TM) 97.0+%, TCI America™
Supplier: TCI America B54801G
Specifications
| AlkylFluor(TM) | |
| C27H36BF5N2 | |
| 1759 | |
| ZXIKPDSPOFQTLH-UHFFFAOYSA-N | |
| 1,3-bis[2,6-di(propan-2-yl)phenyl]-2-fluoroimidazol-1-ium;tetrafluoroborate | |
| 124203184 | |
| ≥97.0% (N) |
| 2043361-32-4 | |
| 1 g | |
| 1,3-Bis(2,6-diisopropylphenyl)-2-fluoroimidazolium Tetrafluoroborate | |
| [B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2F)C3=C(C=CC=C3C(C)C)C(C)C | |
| 494.401 | |
| 494.40 | |
| Crystalline Powder |
Chemical Identifiers
| 2043361-32-4 | |
| 494.401 | |
| 1,3-Bis(2,6-diisopropylphenyl)-2-fluoroimidazolium Tetrafluoroborate | |
| 1,3-bis[2,6-di(propan-2-yl)phenyl]-2-fluoroimidazol-1-ium;tetrafluoroborate |
| C27H36BF5N2 | |
| ZXIKPDSPOFQTLH-UHFFFAOYSA-N | |
| 124203184 | |
| [B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2F)C3=C(C=CC=C3C(C)C)C(C)C |
Safety and Handling
TSCA : No