missing translation for 'onlineSavingsMsg'
Learn More

AlkylFluor(TM) 97.0+%, TCI America™

Catalog No. B5480200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5480200MG 200 mg
B54801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5480200MG Supplier TCI America Supplier No. B5480200MG

Chemical Identifiers

CAS 2043361-32-4
Molecular Formula C27H36BF5N2
Molecular Weight (g/mol) 494.401
InChI Key ZXIKPDSPOFQTLH-UHFFFAOYSA-N
Synonym 1,3-Bis(2,6-diisopropylphenyl)-2-fluoroimidazolium Tetrafluoroborate
PubChem CID 124203184
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-2-fluoroimidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2F)C3=C(C=CC=C3C(C)C)C(C)C

Specifications

Quantity 200 mg
UN Number 1759
Formula Weight 494.40
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material AlkylFluor(TM)