missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkane Standard Mixture, For Performance Tests of GC-systems, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Description | Contains C10 and all even n-alkanes from C20 to C40, 50 mg/L in n-heptane each |
| Density | 0.684 g/mL (at 20°C) |
| Quantity | 2 mL |
| Grade | Analytical Standard |