missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Iodide Solution, For Dissolved Oxygen, Certified, LabChem™

$164.04 - $654.32

Specifications

Color Clear, colorless
Format Liquid
Storage Requirements Room Temperature
Type Alkaline Iodide Solution
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC106401
View Documents
LabChem
LC106401
500 mL Amber Poly
Each for $164.04
Only null left
 
LC106402
View Documents
LabChem
LC106402
1 L Amber Poly
Each for $250.43
Only null left
 
LC106404
View Documents
LabChem
LC106404
4 L Poly Bottle
Each for $654.32
Only null left
 
Description

Description

Specifications

Specifications

Clear, colorless
Liquid
Room Temperature
Alkaline Iodide Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'