missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Iodide Azide Solution II, For Dissolved Oxygen (Concentrations Greater than 15mg/L), LabChem™
Specifications
Specifications
| Color | Clear, colorless |
| Format | Liquid |
| Packaging | Amber Poly |
| Quantity | 1 L |
| Storage Requirements | Room Temperature |
| Type | Alkaline Iodide Azide Solution II |