missing translation for 'onlineSavingsMsg'
Learn More

Alkaline-Iodide-Azide, Pomeroy Formulation, Ricca Chemical

For Dissolved Oxygen (DO) Analysis

$1,055.27

Specifications

Color Clear to turbid
pH Range >13
Format Liquid
Packaging Natural Poly Bottle
Type Alkaline-Iodide-Azide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
53516
View Documents Promotion Details
N/A 500 mL
Each for $1,055.27
Only null left
 
Description

Description

Specifications

Specifications

Clear to turbid
Liquid
Alkaline-Iodide-Azide
>13
Natural Poly Bottle
Videos
SDS
Documents

Documents