missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline-Iodide-Azide, Alsterberg Formulation, Ricca Chemical
For Dissolved Oxygen (DO) Analysis
$251.33 - $1,238.00
Specifications
| Color | Clear colorless |
|---|---|
| pH Range | >13 |
| Format | Liquid |
| Packaging | Natural Poly Bottle |
| Type | Alkaline-Iodide-Azide |
Specifications
| Clear colorless | |
| Liquid | |
| Alkaline-Iodide-Azide |
| >13 | |
| Natural Poly Bottle |