Learn More
Alkali Scientific CellPro™ 2.2 ml 96-Well Deep Well Plate, U-Bottom, Square Well, Sterile, 5/pk, 50/cs
Catalog No. 502336110
Encompass
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately). They are autoclavable and sterilized by gamma irradiation in accordance with ISO 13485.
Catalog No. 502336110 Supplier Alkali Scientific Supplier No. MPD25S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately). They are autoclavable and sterilized by gamma irradiation in accordance with ISO 13485.