Learn More
Alkali Scientific CellPro™ 2.0 ml 96-Well Deep Well Plate, U-Bottom, Round Well, Sterile, 5/pk, 50/cs
Catalog No. 502336047
Encompass
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately). They are autoclavable and E-beam sterilized.
Catalog No. 502336047 Supplier Alkali Scientific Supplier No. MPD35S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately). They are autoclavable and E-beam sterilized.