Learn More
Alkali Scientific CellPro™ 1.0 ml 96-Well Deep Well Plate, U-Bottom, Round Well, 5/pk, 50/cs
Catalog No. 502336034
Encompass
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Catalog No. 502336034 Supplier Alkali Scientific Supplier No. MPD25
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates have black printed alpha-numeric reference grids, are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).