missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Alkali-Cation™ Yeast Transformation Kit

Catalog No. MP112200200
Change view
Click to view available options
No. of Reactions:
250 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP112200200 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112200200 Supplier MP Biomedicals™ Supplier No. 112200200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides all of the reagents required to perform a quick and easy transformation. Alkali-Cation™ Yeast Transformation Kit is easy to use, robust, and its scalability makes it great for the high-efficiency transformation needed in two hours.

  • An efficient, non-electrical method for transforming yeast with linear or plasmid DNA
  • Host cells are made competent by treatment with a Lithium/Cesium Acetate mixture
  • The entire procedure may be completed in less than 90 minutes
  • Routinely provides a transformation efficiency of greater than 104 transformants per μg of DNA
  • Has been successfully used to transform linear DNA into Pichia pastoris for protein expression studies

Specifications

Cell Line S. cerevisiae, S. pombe, S. albicans, many strains
Content And Storage 4°C
Format Kit
Sample Type Yeast Culture
For Use With (Application) Yeast transfomation
No. of Reactions 250 Preps
Quantity 500 μL or 3 to 4 mm colony
Synonym High Efficiency Yeast Transformation Kit

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.