missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow R sodium salt

CAS: 1718-34-9 | C13H8N3NaO5 | 309.213 g/mol

$131.15 - $170.71

Chemical Identifiers

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00067121
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Synonym alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
PubChem CID 44135758
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3870709
View Documents
Thermo Scientific Chemicals
03870709
10 g
Each for $131.15
Only null left
 
AA3870714
View Documents
Thermo Scientific Chemicals
03870714
25 g
Each for $170.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1718-34-9
309.213
HKSANXZSKDUDBD-UHFFFAOYSA-M
44135758
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]
C13H8N3NaO5
MFCD00067121
alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate

Specifications

Brown to Yellow
Powder
C13H8N3NaO5
962365
HKSANXZSKDUDBD-UHFFFAOYSA-M
sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
44135758
Odorless
Alizarin Yellow R sodium salt
1718-34-9
10 g
MFCD00067121
alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]
309.213
309.22
Dark yellow to brown powder
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 217-002-4

RTECSNumber : DH2528600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only