missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow GG, TCI America™

Catalog No. A057825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A057825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A057825G Supplier TCI America Supplier No. A057825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 584-42-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00044746
InChI Key GCGHYYRSTZNXJS-UHFFFAOYSA-M
Synonym metachrome yellow, alizarin yellow gg, benzoic acid, 2-hydroxy-5-3-nitrophenyl azo-, monosodium salt, sodium 2-hydroxy-5-3-nitrophenyl diazenyl benzoate, sodium 2-hydroxy-5-e-2-3-nitrophenyl diazen-1-yl benzoate, sodium 2-hydroxy-5-2-3-nitrophenyl diazen-1-yl benzoate, benzoic acid, 2-hydroxy-5-2-3-nitrophenyl diazenyl-, sodium salt 1:1, sodium 3-3-nitrophenyl hydrazinylidene-6-oxidanylidene-cyclohexa-1,4-diene-1-carboxylate, sodium 3-3-nitrophenyl hydrazinylidene-6-oxo-1-cyclohexa-1,4-dienecarboxylate, sodium 3-2-3-nitrophenyl hydrazin-1-ylidene-6-oxocyclohexa-1,4-diene-1-carboxylate
PubChem CID 44135356
IUPAC Name sodium;3-[(3-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])NN=C2C=CC(=O)C(=C2)C(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 309.21
Physical Form Crystalline Powder
Chemical Name or Material Alizarin Yellow GG
EINECSNumber (5)-2044
RTECSNumber DH2528550
TSCA Yes