missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S sodium salt

Catalog No. p-4407847
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4204006 5 g
AA4204014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4204006 Supplier Thermo Scientific Chemicals Supplier No. 04204006
Only null left

Transition interval: pH 4.6 (yellow) to pH 6.0 (red) | CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

Alizarin Red S sodium salt is used as a chromogenic agent for the colorimetric determination of dothiepin hydrochloride in pharmaceutical formulations. It is also used as an ion-pair reagent for the spectrophotometric assay of fexofenadine in pharmaceuticals. It acts as a colorometric pH indicator in the range of 3.8-5.4. Further, it is used in a biochemical assay to determine the presence of calcific deposition by cells of an osteogenic lineage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym alizarin red s, sodium alizarinesulfonate, mordant red 3, sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, alizarine s, acid red alizarine, sodium alizarin-3-sulfonate, alizarine red s sodium salt, alizarinsulfonate, alizarin s
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Physical Form Powder
Quantity 5 g
Beilstein 4117090
Solubility Information Soluble in water and alcohol.
Formula Weight 342.27
Appearance Various color powder
Chemical Name or Material Alizarin Red S sodium salt
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-981-8
RTECSNumber CB1095300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only