missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin Red S, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | Pouch |
| Color | Brown to Orange |
| pH | 4.5 to 5 |
| Quantity | 25 g |
| Physical Form | Powder |
| Grade | Indicator |
| Chemical Name or Material | Alizarin Red S |
Safety and Handling
| ShelfLife | 60 months |