missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Red S, Reagent Grade, LabChem™

Catalog No. LC105908
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC105908 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC105908 Supplier LabChem Supplier No. LC105908

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Linear Formula C14H7NaO7S
Decomposition Information Sodium oxide Carbon monoxide Carbon dioxide Sulfur compounds
Color Orange/Red
Solubility Information Soluble in water
Quantity 25 g
Physical Form Powder
Formula Weight 342.27
Grade Reagent
Chemical Name or Material Alizarin Red S
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If on skin: Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature