missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Astrol, TCI America™

Catalog No. A060925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A060925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A060925G Supplier TCI America Supplier No. A060925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6408-51-1
Molecular Formula C22H17N2NaO5S
Molecular Weight (g/mol) 444.437
MDL Number MFCD00058945
InChI Key RIJLWEYDGZAVGC-UHFFFAOYSA-M
PubChem CID 23670845
IUPAC Name sodium;5-methyl-2-[[4-(methylamino)-9,10-dioxoanthracen-1-yl]amino]benzenesulfonate
SMILES CC1=CC(=C(C=C1)NC2=C3C(=C(C=C2)NC)C(=O)C4=CC=CC=C4C3=O)S(=O)(=O)[O-].[Na+]

Specifications

Color Black
Quantity 25 g
Formula Weight 444.44
Physical Form Crystalline Powder
Chemical Name or Material Alizarin Astrol
EINECSNumber (5)-1637
TSCA Yes