missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Aliphatic Amine Latex Beads, 2% w/v, 3 μm

Catalog No. A37368
Encompass
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A37368 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A37368 Supplier Molecular Probes™ Supplier No. A37368

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene.

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aliphatic amine latex has a high density of amine groups attached to the terminus of an aliphatic six-carbon spacer arm. This product can be used to covalently couple proteins. Location of the amine at the end of the spacer arm minimizes difficulties with steric hindrance and conformation since the group has a greater freedom to rotate. In addition, spacer arms improve the kinetics of latex agglutination reactions. The particles are stabilized by the positively-charged amine groups under low to neutral pH conditions. Care should be taken not to use them under high pH.

Microspheres Are Available in Many Sizes and Surface Functionalities
• 25 sizes to choose from—0.02 microns to 16 microns
• 20 different surface—for ready conjugation to virtually any material
• Surfactant free—ultraclean
• Various quantities—from milliliter to 500-liter amounts

Key Applications of Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Specifications

Content And Storage Store in refrigerator (2–8°C).
Material Polystyrene
Product Type Latex Beads
Product Line IDC
Quantity 15 mL
Shipping Condition Room Temperature
Surface Modification Aliphatic Amine
Diameter (Metric) 3 μm

For Research Use Only. Not for use in diagnostic procedures.