missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies ali-Q™ 2 Aliquoting Pipet Controller

Catalog No. 10101131
Change view
Click to view available options
Model:
ali-Q 2
ali-Q 2 LS
ali-Q 2 VS
Volume (Metric):
0.3 to 3 mL
0.5 to 5 mL
0.5 to 5 mL (Aliquot)
Type:
Fixed high aliquoting speed
Fixed low aliquoting speed
Variable aliquoting speed
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Volume (Metric) Type
10101131 ali-Q 2 0.5 to 5 mL Fixed high aliquoting speed
10101132 ali-Q 2 LS 0.3 to 3 mL Fixed low aliquoting speed
10101133 ali-Q 2 VS 0.5 to 5 mL (Aliquot) Variable aliquoting speed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10101131 Supplier VistaLab™ Technologies Supplier No. 21000005
Only null left

A 2-in-1 pipettor for Repeat Pipetting and Conventional Pipetting. Faster, highly accurate, and reproducible multi-dispensing without having to watch the lines on your serological pipet. Unique one-touch repeat pipetting button, aliquot volume setting dial, and speed dial (for Variable Speed version only).

  • 3 models: ali-Q 2 (fixed high speed), ali-Q 2 LS (fixed low speed), ali-Q 2 VS (variable speed model)
  • Grey triggers are touch sensitive for all models - speeds only apply to aliquoting (when using purple button)
  • On-board/in-lab Calibration capability, Removable kickstand
  • Highly accurate and reproducible aliquoting with the press of a single button
  • No need to eyeball the meniscus
  • Use any brand and size of serological pipets; optimal performance when using Wobble-not™ pipets
  • Intelligent measuring system allows real-time corrections for varying environmental conditions
  • Accuracy and precision maintained at any dispense angle for more comfortable body positions; especially beneficial under a hood
  • Highly sensitive, touch-control aspirate and dispense buttons
  • Self-standing, like all Ovation™ pipettes, with an ergonomic and comfortable grip

Specifications

Accuracy 2% at 0.5 and 5 mL
Battery Type Rechargeable Lithium Ion
Compatible Tips Any serological pipet
Autoclavable Not Autoclavable
Color Purple
Disposable Reusable
For Use With Pipette
Grip Type Ergonomic
Material Plastic
Model ali-Q 2
Number of Channels 1
Sterility Non-sterile
Type Fixed high aliquoting speed
Volume (Metric) 0.5 to 5 mL
Filter Type 0.22 μM PTFE Membrane Filter
Finger Style Sensitive touch-control aspirate and dispense buttons
For Use With (Application) Repeat Pipetting, Multidispensing, Cell Culture, Filling Plates/Tubes
Includes USB C Charger, Wall Mount
Quantity 1 Ea.
Warranty 1 Year Limited Warranty
Show More Show Less