missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alginic Acid, Sodium Salt, Medium Viscosity, MP Biomedicals
Source: kelp
$817.53 - $817.53
Chemical Identifiers
| CAS | 9005-38-3 |
|---|---|
| Molecular Formula | (C6H7O7)A(C6H7O7)BNa |
| Synonym | Sodium alginate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15472490
|
MP Biomedicals Inc
0215472490 |
500 g |
Each for $817.53
|
|
|||||
Chemical Identifiers
| 9005-38-3 | |
| (C6H7O7)A(C6H7O7)BNa | |
| Sodium alginate |
Specifications
| White to Cream | |
| Powder | |
| >98% | |
| (C6H7O6Na)n | |
| Alginic Acid, Sodium Salt, Medium Viscosity, Viscosity (1% aq. solution): 600-900 cps |
| 9005-38-3 | |
| 500 g | |
| (C6H7O7)A(C6H7O7)BNa | |
| Sodium alginate |
Safety and Handling
Recommended Storage : Room Temperature