missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, Sodium Salt, Medium Viscosity, MP Biomedicals

Catalog No. ICN15472490
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15472490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15472490 Supplier MP Biomedicals Inc Supplier No. 0215472490

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Source: kelp

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
Synonym Sodium alginate

Specifications

Color White to Cream
Physical Form Powder
Quantity 500 g
Assay Percent Range >98%
Linear Formula (C6H7O6Na)n
Chemical Name or Material Alginic Acid, Sodium Salt, Medium Viscosity, Viscosity (1% aq. solution): 600-900 cps
Recommended Storage Room Temperature