missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alginic acid, sodium salt

Catalog No. p-51881
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177770050 5 g Glass bottle
AC177772500 250 g Plastic Bottle
AC177775000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC177770050 Supplier Thermo Scientific Chemicals Supplier No. 177770050
Only null left

CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin, Sodium alginate

Specifications

Quantity 5 g
Packaging Glass bottle
Chemical Name or Material Alginic acid, sodium salt
RTECSNumber AZ5820000
TSCA TSCA

RUO – Research Use Only