missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ Firming Buffer

Catalog No. A1091501
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1091501 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1091501 Supplier Gibco™ Supplier No. A1091501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AlgiMatrix™ Firming Buffer

AlgiMatrix™ Firming Buffer is for use with the AlgiMatrix™ 3D Culture System to facilitate the formation of firm sponges in the 96 well and 24 well plate formats. This buffer allows the users to adjust the firmness of the sponge to meet specific cell and experimental requirements. Should be ordered as a separate component.

Specifications

Chemical Name or Material Firming Buffer
For Use With (Equipment) 3D Culture System to facilitate the formation of firm sponges in the 96 well and 24 well plate formats
Physical Form Liquid
Product Line Algimatrix
Quantity 50 mL
Sterility Sterile
Grade Cell Culture Grade
How stable is AlgiMatrix Matrix at room temperature without degradation?

AlgiMatrix Matrix is stable for 12 months unopened. Once opened, the entire plate needs to be used.

Can other cell viability reagents be used with AlgiMatrix Matrix?

Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.

How important is calcium in the AlgiMatrix firming and dissolving buffers?

The firming buffer works by adding calcium to the sponge, making it stiffer. The dissolving buffer works by removing calcium from the sponge, making it softer.

Is there any assay variability when using AlgiMatrix Matrix?

3D cell culture with AlgiMatrix Matrix approaches a more physiological state in terms of cell growth and behavior, but along with that comes the higher variability typically associated with more complex systems. You may need to do more replicates or more controls.

Can I use AlgiMatrix Matrix for primary cells?

Some primary cells (with the exception of stem cells) need to adhere in order to expand. As cells won't adhere to AlgiMatrix Matrix, these cells can still be cultured, but will tend to differentiate rather than expand. We recommend that you plate at a high cell density for primary cells.

What is the depth of the AlgiMatrix Matrix sponge? Will the sponge enlarge after addition of media?

The height of the sponge is ~3-4 mm dry. When the sponge is hydrated, its volume is essentially the same - it doesn't swell and increase in size, in fact it becomes slightly bit smaller as it essentially goes back into a hydrogel consistency, especially if the inoculated sponges are centrifuged briefly (as suggested to force cells more into the interior and also to spread the sponge uniformly over the bottom of the well).

Since normal cells and cancer cells form spheroids in AlgiMatrix Matrix, how do you distinguish normal from cancer cells in a traditional sense? What is the main difference between AlgiMatrix Matrix and soft-agar in this regard?

Alginate, from which AlgiMatrix Matrix is made, presents a very hydrophilic surface, and lipid-containing cell membranes and even molecules do not adhere. Normal cells will aggregate to form spheroids and differentiate into structures in AlgiMatrix Matrix, but expansion will be limited since most normal cells need to adhere to increase in number. Cancer cells are able to expand without attachment and do so in AlgiMatrix Matrix (one exception to this is embryonic stem cells, which can expand as embryoid bodies in AlgiMatrix Matrix).

However, here is one major difference between soft-agar and AlgiMatrix Matrix: soft-agar is a hydrogel and AlgiMatrix Matrix is a sponge with relatively large pore sizes, and cells that do grow will be able to do so easier and faster in AlgiMatrix Matrix than when encased in a hydrogel (soft-agar). So a traditional way to tell cancer cells from normal cells is to observe the culture for expansion; cancer cells (example HepG2 hepatocarcinoma) will form expanding spheroids while normal cells will form spheroids that do not increase in size or number.

The significance of collagen is that it is just like any natural structure in the body, which makes it less "foreign" to the human body for potential transplant research applications. How about AlgiMatrix Matrix?

The recent more highly purified forms of alginate do not generate a sensitivity reaction as is the case with less pure versions. Alginate has been used for some years to encapsulate, for instance, B-cells of the pancreas for in vivo transplantation (diabetes research) and it does not get rejected.

If AlgiMatrix Matrix is extracted from brown seaweed, how can it be pharmaceutical raw material standard and also without excessive lot-to-lot variation?

Our supplier has been producing pharmaceutical-grade alginate and hyaluroinic acid for years. Although these are natural products, by specifying among other things (in the case of alginate) harvest location, water temperature, which part of the seaweed to harvest, use of stringent purification SOPs, and rigorous QC criteria, a very standard product can be obtained.

Can AlgiMatrix Matrix be used for metastasis studies/invasion studies?

Invasion studies measure the ability of cells to penetrate through a matrix, simulating a metastatic cells activity in vivo. AlgiMatrix Matrix contains only alginate (brown seaweed) with no in vivo-like molecules in the undecorated structure, so invasion may not be a very relevant use for AlgiMatrix Matrix.

What are the possible downstream analyses for 3D cell culture? For instance if someone would like to do immunofluorescence/immunohistochemistry staining, how can this be done with a 3D structure? Can the normal fluorescence/light microscope handle the 3D structure staining?

Standard immunofluorescence testing can be performed on 3D structures. The staining can be done with either intact spheroids within the AlgiMatrix Matrix or you can dissolve the AlgiMatrix Matrix using Versene or 55 mM tri-sodium citrate dihydrate and stain the spheroid alone. You can further dissociate the spheroid into individual cells using TrypLE reagent and stain as dissociated cells.

What growth conditions do you recommend when using the AlgiMatrix 3D Culture System for umbilical cord stem cells?

For umbilical cord stem cells, you might want to try a relatively high inoculation density of 100,000-300,000 cells per sponge with daily or every-other-day re-feeding of cultures, as the medium turns “yellow”.

What are the optimal growth conditions when using the AlgiMatrix Matrix 3D Culture System?

This scaffold is great for forming 3D spheroids that allow maximum cell to cell interaction. Established cell lines expand by increasing size of spheroids up to ~100 cells or so depending upon the cell type. Both primary and established cells will differentiate in a 3-dimensional format over 1-2 weeks of culture.

Are there references or protocols that describe how to culture and expand MSCs, HUVECs, and HDMECS in the 3D AlgiMatrix Matrix sponge?

Please see the following references specifically for stem cells:

- Methods Enzymol 420:303 (2006)
- Biotechnol Bioeng 88:313 (2004)

When staining cells or spheroids in the sponge, how problematic is background staining due to AlgiMatrix Matrix?

There is virtually no background staining with alginate. The same surface characteristic that prevents cells from adhering also makes it very difficult for molecules such as fluoresceinated antibodies to adhere, thus essentially no background staining is observed.

Can AlgiMatrix Matrix be used for culturing mouse ES cells?

While we have not tested this in-house, the common belief is that if the alginate scaffolds support proliferation of human ES without feeder cells, they should also support mouse ES. Both mouse and human form embryoid bodies, and seeding within the macro-porous alginate scaffolds have been found very advantageous for these structures, in terms of proliferation and differentiation.

Can I subculture cells in AlgiMatrix Matrix after growing cells for 3 to 4 days and at certain density?

Yes, start by dissolving the matrix using calcium binding solution, Versene, or sodium citrate. The remaining spheroids can be dissociated with TrypLE reagent, counted, and expanded into other culture vessels or into new AlgiMatrix Matrix sponges. These steps are illustrated in the instruction manual that is supplied with each kit.

How long does spheroid formation take in the Algimatrix 3D Culture System?

Spheroid formation requires about 5 to 7 days once inoculated. Air bubbles will be observed at the time of inoculation. This will dissipate within several days by the cells consuming the oxygen.

What level of force can be used when re-feeding cells while using the Algimatrix 3D Culture System?

Add medium gently at an angle. Be careful not to impel the matrix when re-feeding.

Is there any data showing penetration and percent cell population, when using the Algimatrix 3D Culture System?

We have shown the following data:

- Focusing on cells in culture shows evidence of inoculation.
- Paraffin embedding and sectioning shows that cells are inoculated throughout the sponge.


For Research Use Only. Not for use in diagnostic procedures.