missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AlgiMatrix™ 6-Well, 3D Culture System, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
$580.00 - $2,245.00
Specifications
| Lid | With Lid |
|---|---|
| Product Line | Algimatrix |
| Color | Clear |
| Product Type | Microplate |
| No. of Wells | 6 |
Description
AlgiMatrix™ 3D Culture System 6 well plate is a user-friendly, animal –free bioscaffold available for the development of high-fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ sponges are formulated using pharmaceutical-grade raw material from brown seaweed. Its pure, chemically defined, and nontoxic macro porous structure improved lot-to-lot consistency, superior cell loading and excellent nutrient delivery, without damage to cells. The AlgiMatrix™ sponge is biodegradable and is stable at room temperature.The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
Specifications
| With Lid | |
| Clear | |
| 6 | |
| AlgiMatrix 3D Culture System | |
| Cell Culture Grade |
| Algimatrix | |
| Microplate | |
| Sterile | |
| Flat Bottom (F-well) | |
| Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.