missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022943
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022943 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022943 Supplier Merck Emd Millipore Supplier No. BP847

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 81403-68-1
Quantity 25 mg
Molecular Formula C19H27N5O4 · HCl
Linear Formula C19H27N5O4 · HCl
MDL Number MFCD00879135
Synonym Alfuzosin hydrochloride; N-(3-((4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino)propyl)tetrahydro-2-furancarboxamide hydrochloride; Uroxatral hydrochloride
Formula Weight 425.91
Grade BP