missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfuzosin Hydrochloride 98.0+%, TCI America™
$88.92 - $397.60
Chemical Identifiers
| CAS | 81403-68-1 |
|---|---|
| Molecular Formula | C19H28ClN5O4 |
| Molecular Weight (g/mol) | 425.914 |
| MDL Number | MFCD00879135 |
| InChI Key | YTNKWDJILNVLGX-UHFFFAOYSA-N |
| Synonym | alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od |
| PubChem CID | 71764 |
| ChEBI | CHEBI:32286 |
| IUPAC Name | N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride |
| SMILES | CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2591100MG
|
TCI America
A2591100MG |
100 mg |
Each for $88.92
|
|
|||||
|
A25911G
|
TCI America
A25911G |
1 g |
Each for $397.60
|
|
|||||
Chemical Identifiers
| 81403-68-1 | |
| 425.914 | |
| YTNKWDJILNVLGX-UHFFFAOYSA-N | |
| 71764 | |
| N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride |
| C19H28ClN5O4 | |
| MFCD00879135 | |
| alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od | |
| CHEBI:32286 | |
| CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl |
Specifications
| 229°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00879135 | |
| YTNKWDJILNVLGX-UHFFFAOYSA-N | |
| N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride | |
| 71764 | |
| 425.91 | |
| Alfuzosin Hydrochloride |
| 81403-68-1 | |
| 100 mg | |
| C19H28ClN5O4 | |
| alfuzosin hydrochloride, uroxatral, alfuzosin hcl, xatral, urion, alfoten, alfetim, xatral retard, xatral od | |
| CN(CCCNC(=O)C1CCCO1)C2=NC3=CC(=C(C=C3C(=N2)N)OC)OC.Cl | |
| 425.914 | |
| CHEBI:32286 | |
| ≥98.0% (HPLC) |
Safety and Handling
RTECSNumber : LT9965475
TSCA : No