missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 647-Biotin Control Peptide
Specifications
Specifications
| Product Line | Alexa Fluor |
| Color | Far-Red |
| Content And Storage | 100 nM Alexa Fluor 647-Biotin Control peptide in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl (HEPES buffered saline). Store at -20°C and protect from light. |
| Format | Frozen |
| Label or Dye | Alexa Fluor™ 647 |
| Label Type | Alexa Fluor Dyes |
| Quantity | 0.4 mL |
| Research Category | Drug Discovery, Immunology |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.