missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 555 Azide, Triethylammonium Salt

Catalog No. a20012
Encompass
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20012 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20012 Supplier Molecular Probes™ Supplier No. A20012
Only null left

Alexa Fluor™ 555 azide, triethylammonium salt

The orange-fluorescent Alexa Fluor™ 555 azide is designed to react with alkyne-containing molecules via the fast, selective and extremely efficient copper-catalyzed click reaction.

Specifications

Quantity 0.5 mg
Product Type Azide
Content And Storage Store at -20°C, dessicate and protect from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Amine, Azide
Label or Dye Alexa Fluor™ 555
Label Type Alexa Fluor
Color Orange
Product Line Alexa Fluor
Emission 565
Excitation 555
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.