missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 555 alkyne, Triethylammonium Salt

Catalog No. a20013
Encompass
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20013 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20013 Supplier Molecular Probes™ Supplier No. A20013
Only null left

Alexa Fluor™ 555 alkyne, triethylammonium salt

The orange-fluorescent Alexa Fluor™ 555 alkyne is designed to react with azide-containing molecules via the fast, selective and extremely efficient copper-catalyzed click reaction.

Specifications

Quantity 0.5 mg
Product Type Alkyne
Content And Storage Store at -20°C, dessicate and protect from light.
Chemical Reactivity Azide
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Alkyne
Label or Dye Alexa Fluor™ 555
Label Type Alexa Fluor
Color Orange
Product Line Alexa Fluor
Emission 565
Excitation 555
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.