missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies

Catalog No. A11054
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11054 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11054 Supplier Invitrogen™ Supplier No. A11054
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies

For sensitive detection of mouse primary antibodies, try our Alexa Fluor™ 488 Signal-Amplification Kit, which contains Alexa Fluor 488 rabbit anti—mouse IgG and Alexa Fluor 488 goat anti—rabbit IgG conjugates. This kit takes advantage of the intense, photostable fluorescence of Alexa Fluor 488 antibody conjugates and can be used to amplify the signals derived from mouse-derived antibodies. The kit contains a detailed protocol and sufficient reagents for ∼60 assays by flow cytometry or imaging. For added convenience, the kit is designed for use with filters and settings appropriate for fluorescein.

Specifications

Product Line Alexa Fluor
Quantity 1 kit
Shipping Condition Room Temperature
Label or Dye Alexa Fluor™ 488
Content And Storage Store in refrigerator (2–8°C) and protect from light.

For Research Use Only. Not for use in diagnostic procedures.