missing translation for 'onlineSavingsMsg'
Learn More

Abbott™ ID NOW Influenza A and B Control Swab Kit

Catalog No. 23046619
Encompass
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23046619 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23046619 Supplier Abbott™ Supplier No. 425080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides molecular results in less than 15 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.

Transforming near-patient testing

  • Unique isothermal amplification technology giving you molecular results, faster than ever before
Improving patient outcomes
  • Giving you the confidence to make effective clinical decisions sooner
Optimizing operational efficiency
  • Enables prompt initiation of infection control measures
  • Connected results direct to your network
  • Facilitating effective patient management
Enabling effective Antimicrobial Stewardship
  • Molecular sensitivity in an actionable timeframe allows the prudent use of antibiotics and antivirals
A positive impact on your facility
  • Reliable near-patient testing reduces overall healthcare costs
Confidence at the point of care
  • Intuitive and reliable, allowing you to respond quickly and accurately in any setting
  • Simple and easy to use molecular diagnostic test
  • Sample type nasal swab
  • Requires only minimal training
  • Small footprint can be used in any healthcare setting
  • Streamlined workflow connectivity can be provided to your network
  • Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens

 

Specifications

Type Abbott ID NOW Influenza A and B Control Swab Kit
For Use With (Application) Influenza testing
Quantity 20/Pk.