missing translation for 'onlineSavingsMsg'
Learn More

Abbott™ BinaxNOW™ Streptococcus pneumoniae Antigen Card Test Kit

Catalog No. BNX710000
Encompass
Change view
Click to view available options
Quantity:
22 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BNX710000 22 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BNX710000 Supplier Abbott™ Supplier No. 710000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BinaxNOW™ S. pneumoniae Antigen Card is a rapid assay for the qualitative detection of S. pneumoniae antigen in the urine of patients with pneumonia and in the cerebral spinal fluid (CSF) of patients with meningitis. It is intended to aid, in conjunction with culture and other methods, in the diagnosis of both pneumococcal pneumonia and pneumococcal meningitis.

  • CLIA Complexity: Moderate (Urine, urinary antigen and CSF)
  • Rapid, immunochromatographic assay used in conjunction with culture and other methods to aid in the diagnosis of both pneumococcal pneumonia and pneumococcal meningitis
  • Results in 15 minutes
  • Overall accuracy: 93% urine; 98% CSF
  • For in vitro diagnostic use

Order Info

For control swabs to be used with this test, see Cat. No. BNX710010.

Specifications

Content And Storage Room Temperature
Detectable Analytes S. pneumoniae Antigen
Sample Type CSF, Urine
Type S. pneumoniae Antigen Card
Includes Test devices, reagent A, sample swabs, positive and negative control
Quantity 22 Tests/Kit
Test Time 15 min.