missing translation for 'onlineSavingsMsg'
Learn More

Alendronic Acid 98.0+%, TCI America™

Catalog No. A21205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21205G 5 g
A212025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21205G Supplier TCI America Supplier No. A21205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66376-36-1
Molecular Formula C4H13NO7P2
Molecular Weight (g/mol) 249.096
MDL Number MFCD00868112
InChI Key OGSPWJRAVKPPFI-UHFFFAOYSA-N
Synonym 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid
PubChem CID 2088
ChEBI CHEBI:2567
IUPAC Name (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid
SMILES C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 249.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Alendronic Acid
RTECSNumber SZ6521833
TSCA No