missing translation for 'onlineSavingsMsg'
Learn More

Alendronate sodium trihydrate, 97%

Catalog No. AAJ6139703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6139703 1 g
AAJ6139706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6139703 Supplier Thermo Scientific Chemicals Supplier No. J6139703

CAS: 121268-17-5 | C4H18NNaO10P2 | 325.122 g/mol

Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.

Solubility
Soluble in water at 10mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 121268-17-5
Molecular Formula C4H18NNaO10P2
Molecular Weight (g/mol) 325.122
MDL Number MFCD01748233
InChI Key DCSBSVSZJRSITC-UHFFFAOYSA-M
Synonym alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan
PubChem CID 23681107
ChEBI CHEBI:2566
IUPAC Name sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate
SMILES C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+]

Specifications

Color White
Quantity 1 g
Merck Index 14,229
Solubility Information Soluble in water at 10mg/ml
Formula Weight 325.12 (271.08 Anhydrous)
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Alendronate sodium trihydrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber SZ6523500
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only