Learn More
MilliporeSigma™ Supelco™ Aldrich™ TLC Developing Tank - Cylindrical Glass Tank
Aldrich™ TLC developing tanks are used to develop TLC plates.
Supplier: MilliporeSigma™ Supelco™ Z2439066X1EA
Description
Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.Chemical Identifiers
| Glass | |
| TLC | |
| Aldrich | |
| 6.5 cm |
Specifications
| Glass | |
| TLC | |
| 10.5 cm |
| 6 x 1/Pk. | |
| Aldrich | |
| 6.5 cm |
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC