Learn More
MilliporeSigma™ Supelco™ Aldrich™ Rectangular TLC Developing Tanks, Complete
TLC Developing Tanks come complete with the developing tank and lid.
$273.16 - $333.71
Specifications
Material Glass
For Use With (Application) TLC
Includes Developing tank and lid
Product Line Aldrich
View More Specs
Products 2
Catalog Number Mfr. No. Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
111010665
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z1462261EA
10.8 cm 12.1 cm 8.3 cm
Each for $273.16
Only null left
111010462
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z1261951EA
26.5 cm 27 cm 7 cm
Each for $333.71
Only null left
Description
Description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results.
Specifications
Specifications
Glass
TLC
Developing tank and lid
Aldrich
Videos
SDS
Documents
Documents
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC