missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Solution, 100 μg/mL in Acetonitrile, Ampule of 1 mL, Certified Reference Material, Cerilliant™
Description
- Aldosterone is a hormone in the mineralocorticoid family which stimulates sodium transport across cell membranes and assists in the maintenance of blood pressure and blood volume.
- Aldosterone levels in serum/plasma and urine are measured by LC-MS/MS for investigation of primary or secondary aldosteronism ranging from adrenal cortical hyperplasia to renovascular disease and Bartter's syndrome.
Specifications
Specifications
| Chemical Name or Material | Aldosterone |
| Packaging | Ampule |
| CAS | 52-39-1 |
| Molecular Formula | C21H28O5 |
| Concentration | 100 μg/mL |
| For Use With (Equipment) | Liquid Chromatography (LC); Gas Chromatography (GC) |
| Solution Type | Standard Solution |
| Type | Certified Reference Material |
| Industry Type | Clinical Testing |
| Molecular Weight (g/mol) | 360.44 |
| Show More |