missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Aldehyde/Sulfate Latex Beads, 4% w/v, 4 μm

Catalog No. A37304
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A37304 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A37304 Supplier Molecular Probes™ Supplier No. A37304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene.

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aldehyde/sulfate latex contains an abundance of aldehyde groups grafted to the surface of the polymer particle. The high density of aldehyde groups enables facile coupling of proteins and other materials to the latex particles in a one-step process. The sulfate charge groups on the microspheres enable retention of stability during the covalent coupling process. Due to the high density of polar groups, this latex has a hydrophilic surface.

Microspheres Are Available in Many Sizes and Surface Functionalities
• 25 sizes to choose from—0.02 microns to 16 microns
• 20 different surface—for ready conjugation to virtually any material
• Surfactant free—ultraclean
• Various quantities—from milliliter to 500-liter amounts

Key Applications of Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Specifications

Content And Storage Store in refrigerator (2–8°C).
For Use With (Application) Flow Cytometry
Product Type Latex Beads
Product Line IDC
Quantity 15 mL
Shipping Condition Room Temperature
Surface Modification Aldehyde Sulfate

For Research Use Only. Not for use in diagnostic procedures.