missing translation for 'onlineSavingsMsg'
Learn More

Alconox™ Liquinox™ Critical Cleaning Liquid Detergent
GREENER_CHOICE

Catalog No. 16000125
Change view
Click to view available options
Volume (English):
1 qt.
1 gal.
5 gal.
15 gal.
55 gal.
Container Type:
Case
Drum
Jerrican
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16000125 1 qt. Case
16000127 5 gal. Jerrican
16000126 1 gal. Case
0432215C 15 gal. Drum
16000129 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 16000125 Supplier Alconox™ Supplier No. ALC1232
Edge

Concentrated, mild, anionic detergent for manual and ultrasonic cleaning

This concentrated, mild, anionic detergent is suitable for manual and ultrasonic cleaning.

Liquinox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, sodium xylene sulfonate, alkanolamide, and ethoxylated alcohol.

 

  • Concentrated
  • Phosphate free, biodegradable and readily disposable
  • Completely soluble in hard and soft water
  • Replaces corrosive acids and hazardous solvents
  • Dilute 1:100 (dilute 10mL in 1L of water)
  • Free rinsing—gives reliable results without interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants

 

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors. Passes inhibitory residue test for water analysis. Used for phosphate sensitive analysis ware.

Used to remove:
Soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemicals and solvents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life: 2 years from date of manufacture

Order Info

3.8L (1 gal.) makes 378.5L (100 gal.) cleaning solution.

Specifications

Product Type Critical Cleaning Liquid Detergent
Form Liquid
pH 8.5
Appearance Pale Yellow
Container Type Case
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Clean healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware
Quantity 12/Cs.
Volume (English) 1 qt.
Volume (Metric) 0.95 L