missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alcian Blue 8GX, certified

CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol

Supplier:  Thermo Scientific Chemicals 400460250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC400460250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33864-99-2
1298.86
XRWQTDIELZDCPS-UHFFFAOYSA-J
17749099
[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
C56H68Cl4CuN16S4
MFCD00010720
Ingrain Blue 1
(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride

Specifications

Alcian Blue 8GX
33864-99-2
C56H68Cl4CuN16S4
15, 212
Solubility in water: moderately soluble. Other solubilities: soluble in ethanol, cellosolve, ethylene glycol, practically insoluble in xylene
[Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C
(9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
17749099
Glass bottle
25 g
148.0°C
50% min.
MFCD00010720
Ingrain Blue 1
XRWQTDIELZDCPS-UHFFFAOYSA-J
Authentic
1298.86
1298.88
Purple
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions