missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alcian Blue 8GX, certified
CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Supplier: Thermo Scientific Chemicals 400460100
| Quantity | 10 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 33864-99-2 | |
| 1298.86 | |
| XRWQTDIELZDCPS-UHFFFAOYSA-J | |
| 17749099 | |
| [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C |
| C56H68Cl4CuN16S4 | |
| MFCD00010720 | |
| Ingrain Blue 1 | |
| (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride |
Specifications
| Alcian Blue 8GX | |
| 33864-99-2 | |
| C56H68Cl4CuN16S4 | |
| 15, 212 | |
| Solubility in water: moderately soluble. Other solubilities: soluble in ethanol, cellosolve, ethylene glycol, practically insoluble in xylene | |
| [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C | |
| (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride | |
| 17749099 | |
| Glass bottle | |
| 10 g |
| 148.0°C | |
| 50% min. | |
| MFCD00010720 | |
| Ingrain Blue 1 | |
| XRWQTDIELZDCPS-UHFFFAOYSA-J | |
| Authentic | |
| 1298.86 | |
| 1298.88 | |
| Purple | |
| Crystalline Powder |