Learn More
Thermo Scientific Chemicals Alcian Blue 8GX
CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Supplier: Thermo Scientific Chemicals J6012222
| Quantity | 100 g |
|---|
Description
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAlcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
Solubility
Soluble in water, ethanol, ethylene glycol and 2-ethoxyethanol. Insoluble in xylene.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 33864-99-2 | |
| 1298.86 | |
| XRWQTDIELZDCPS-UHFFFAOYSA-J | |
| 17749099 | |
| [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C |
| C56H68Cl4CuN16S4 | |
| MFCD00010720 | |
| Ingrain blue 1; C.I. 74240 | |
| (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride |
Specifications
| Alcian Blue 8GX | |
| 33864-99-2 | |
| MFCD00010720 | |
| Ingrain blue 1; C.I. 74240 | |
| XRWQTDIELZDCPS-UHFFFAOYSA-J | |
| (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride | |
| 17749099 | |
| Blue | |
| Powder |
| 148°C | |
| C56H68Cl4CuN16S4 | |
| 14,218 | |
| Soluble in water,ethanol,ethylene glycol and 2-ethoxyethanol; Insoluble in xylene. | |
| [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C | |
| 1298.86 | |
| 1298.88 | |
| 100 g |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 251-705-7
TSCA : Yes
Recommended Storage : Ambient temperatures