Learn More
BINDER™ Alarm Output
Description
- Alarm output
- Zero-voltage
- Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
- Acoustic signal that can be switched off
Specifications
Specifications
| For Use With (Equipment) | KBF Series |
| Material | Copper |
| Type | Analog Output |