missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine-tert-butylester hydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H15NO2·ClH
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
SMILES CC(C)(C)OC(=O)CCN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434650010
SDS View Documents
Thermo Scientific Chemicals
434650010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434650050
SDS View Documents
Thermo Scientific Chemicals
434650050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

58620-93-2
181.66
DOMTZTVJNZKUNX-UHFFFAOYSA-N
16218890
CC(C)(C)OC(=O)CCN.Cl
C7H15NO2·ClH
MFCD00058258
tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride
tert-butyl 3-aminopropanoate;hydrochloride

Specifications

58620-93-2
100.0
Authentic
Glass bottle
MFCD00058258
tert-butyl 3-aminopropanoate hydrochloride, tert-butyl 3-aminopropanoate-hcl, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, beta-alanine t-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-?-ala-otbu.hcl, a-ala-otbu.hcl, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride
CC(C)(C)OC(=O)CCN.Cl
181.66
181.66
β-Alanine-tert-butylester hydrochloride
97.5
45.1 to 47.4 % C 7.5 to 7.9 % N
98%
C7H15NO2·ClH
1g
DOMTZTVJNZKUNX-UHFFFAOYSA-N
tert-butyl 3-aminopropanoate;hydrochloride
16218890
98%
Videos
SDS
Documents

Documents

RUO – Research Use Only