missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine-tert-butylester hydrochloride, 98%

Catalog No. AC434650010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434650010 1 g Glass Bottle
AC434650050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434650010 Supplier Thermo Scientific Chemicals Supplier No. 434650010
Only null left

CAS: 58620-93-2 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CCN

Specifications

Melting Point 167.8°C to 168.5°C
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Elemental Analysis 45.1 to 47.4 % C 7.5 to 7.9 % N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 181.66
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material β-Alanine-tert-butylester hydrochloride
Show More Show Less

RUO – Research Use Only