missing translation for 'onlineSavingsMsg'
Learn More

Alamethicin

Catalog No. p-7057417
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63829MCR 1 mg
AAJ63829LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63829MCR Supplier Thermo Scientific Chemicals Supplier No. J63829MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A channel-forming ionophore that activates membrane enzymes | CAS: 59588-86-2 | C92H150N22O25 | 1964.341 g/mol

Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

Solubility
Soluble in DMSO and ethanol.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 59588-86-2
Molecular Formula C92H150N22O25
Molecular Weight (g/mol) 1964.341
MDL Number MFCD00151517
InChI Key LGHSQOCGTJHDIL-SLKIUSOBSA-N
Synonym U-22324
PubChem CID 53229968
SMILES CC(C)CC(C(=O)NC(C)(C)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CC2=CC=CC=C2)CO)NC(=O)CNC(=O)C(C)(C)NC(=O)C(C(C)C)NC(=O)C(C)(C)NC(=O)C(CCC(=O)N)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C3CCCN3C

Specifications

Type Alamethicin
Physical Form Powder
Melting Point 267°C to 270°C
Color White
Quantity 1 mg
UN Number UN3462
Solubility Information Soluble in DMSO and ethanol.
Formula Weight 1964.4
Chemical Name or Material Alamethicin
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECSNumber AY1900000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only