missing translation for 'onlineSavingsMsg'
Learn More

Alachlor, Restek

Catalog No. 06719837
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719837 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719837 Supplier Chromatographic Supplier No. RK 32204
Only null left

Chemical Identifiers

CAS 15972-60-8
Molecular Formula C14H20ClNO2
Molecular Weight (g/mol) 269.769
InChI Key XCSGPAVHZFQHGE-UHFFFAOYSA-N
PubChem CID 2078
ChEBI CHEBI:2533
IUPAC Name 2-chloro-N-(2,6-diethylphenyl)-N-(methoxymethyl)acetamide
SMILES CCC1=C(C(=CC=C1)CC)N(COC)C(=O)CCl

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Methanol
Chemical Name or Material Alachlor, Pesticide Mix