missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Round Disc With Hole, O.D.: 80mm, Thickness.: 8mm, Hole Dia.: 10mm, Thermo Scientific™

Catalog No. AA33147KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33147KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33147KF Supplier Thermo Scientific Chemicals Supplier No. 33147KF

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Al-23 Round Disc With Hole
Quantity 1 Pc.
Form Disc